Skip to Content
Merck
All Photos(2)

Documents

C63006

Sigma-Aldrich

2-Chlorophenothiazine

97%

Synonym(s):

2-Chlorothiodiphenylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8ClNS
CAS Number:
Molecular Weight:
233.72
Beilstein:
176587
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

crystals

mp

195-200 °C (lit.)

SMILES string

Clc1ccc2Sc3ccccc3Nc2c1

InChI

1S/C12H8ClNS/c13-8-5-6-12-10(7-8)14-9-3-1-2-4-11(9)15-12/h1-7,14H

InChI key

KFZGLJSYQXZIGP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Helge Prinz et al.
Journal of medicinal chemistry, 54(12), 4247-4263 (2011-05-14)
A total of 53 N-benzoylated phenoxazines and phenothiazines, including their S-oxidized analogues, were synthesized and evaluated for antiproliferative activity, interaction with tubulin, and cell cycle effects. Potent inhibitors of multiple cancer cell lines emerged with the 10-(4-methoxybenzoyl)-10H-phenoxazine-3-carbonitrile (33b, IC(50) values

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service