C49607
4-Chloromercuribenzoic acid
Synonym(s):
4-(Chloromercurio)benzoic acid, 4-(Hydroxymercuri)benzoic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
ClHgC6H4CO2H
CAS Number:
Molecular Weight:
357.16
Beilstein:
3662892
EC Number:
MDL number:
UNSPSC Code:
12352103
Recommended Products
description
cysteine active site modifier
mp
287 °C (dec.) (lit.)
SMILES string
OC(=O)c1ccc([Hg]Cl)cc1
InChI
1S/C7H5O2.ClH.Hg/c8-7(9)6-4-2-1-3-5-6;;/h2-5H,(H,8,9);1H;/q;;+1/p-1
InChI key
YFZOUMNUDGGHIW-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service