Skip to Content
Merck
All Photos(1)

Key Documents

C38907

Sigma-Aldrich

4-Chloro-3,5-dinitrobenzoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H2(NO2)2CO2H
CAS Number:
Molecular Weight:
246.56
Beilstein:
668253
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

crystals

mp

159-162 °C (lit.)

SMILES string

OC(=O)c1cc(c(Cl)c(c1)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C7H3ClN2O6/c8-6-4(9(13)14)1-3(7(11)12)2-5(6)10(15)16/h1-2H,(H,11,12)

InChI key

PCTFIHOVQYYAMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A A Jacobs et al.
Biochimica et biophysica acta, 872(1-2), 92-97 (1986-07-25)
Modification of lysine residues with 4-chloro-3,5-dinitrobenzoate results in the loss of the binding capacity of K99 fibrillae to horse erythrocytes (Jacobs, A.A.C., van Mechelen, J.R. and de Graaf, F.K. (1985) Biochim. Biophys. Acta 832, 148-155). In the present study we
J Thiele et al.
Applied and environmental microbiology, 54(5), 1199-1202 (1988-05-01)
4-Chlorobenzoate dehalogenase from Pseudomonas sp. strain CBS3 converted 4-chloro-3,5-dinitrobenzoate to 3,5-dinitro-4-hydroxybenzoate and 1-chloro-2,4-dinitrobenzene to 2,4-dinitrophenol. The activities were 0.13 mU/mg of protein for 4-chloro-3,5-dinitrobenzoate and 0.16 mU/mg of protein for 1-chloro-2,4-dinitrobenzene compared with 0.5 mU/mg of protein for 4-chlorobenzoate.
L S Chang et al.
Biochemistry and molecular biology international, 33(6), 1207-1213 (1994-08-01)
Phospholipase A2 (PLA2) from Naja naja atra snake venom was modified with 4-chloro-3,5-dinitrobenzoate, and one major carboxydinitrophenylated (CDNP) PLA2 was separated by high performance liquid chromato-graphy. CDNP-PLA2 contained only one CDNP group on Lys-6 and showed a 93% drop in
Jorge L Lopez et al.
Environmental toxicology and chemistry, 23(5), 1129-1135 (2004-06-08)
Physicochemical characterization of hazardous compounds often is required for the development of structure-reactivity correlations. Physical, chemical, and toxicological properties of target pollutants require determination for an efficient application of wastewater treatments. In the present work, we chose a chloro-nitro-aromatic derivative
E L Gross et al.
Biochimica et biophysica acta, 1016(1), 107-114 (1990-03-15)
Chemical modification of plastocyanin was carried out using 4-chloro-3,5-dinitrobenzoic acid, which has the effect of replacing positive charges on amino groups with negatively charged carboxyl groups. Four singly-modified forms were obtained which were separated using anion exchange FPLC. The four

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service