Skip to Content
Merck
All Photos(1)

Key Documents

C103403

Sigma-Aldrich

1-Cyclohexenylacetonitrile

92%

Synonym(s):

1-Cyclohexene-1-acetonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
R$591.00

R$591.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
R$591.00

About This Item

Linear Formula:
C6H9CH2CN
CAS Number:
Molecular Weight:
121.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$591.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

92%

form

liquid

refractive index

n20/D 1.478 (lit.)

bp

144 °C/90 mmHg (lit.)

density

0.947 g/mL at 25 °C (lit.)

SMILES string

N#CCC1=CCCCC1

InChI

1S/C8H11N/c9-7-6-8-4-2-1-3-5-8/h4H,1-3,5-6H2

InChI key

OYEXEQFKIPJKJK-UHFFFAOYSA-N

Other Notes

remainder cyclohexylideneacetonitrile

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service