Skip to Content
Merck
All Photos(1)

Key Documents

C102601

Sigma-Aldrich

Cyclohexene sulfide

85%, technical grade

Synonym(s):

7-Thiabicyclo[4.1.0]heptane

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
R$1,117.00

R$1,117.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
R$1,117.00

About This Item

Empirical Formula (Hill Notation):
C6H10S
CAS Number:
Molecular Weight:
114.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$1,117.00


Check Cart for Availability

Request a Bulk Order

grade

technical grade

Quality Level

Assay

85%

form

liquid

refractive index

n20/D 1.527 (lit.)

bp

55-58 °C/12 mmHg (lit.)

storage temp.

2-8°C

SMILES string

C1CCC2SC2C1

InChI

1S/C6H10S/c1-2-4-6-5(3-1)7-6/h5-6H,1-4H2

InChI key

PQWJNIJNYRPOAA-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

122.0 °F - closed cup

Flash Point(C)

50 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service