Key Documents
BR94910
BRAND® standard ground joint funnel
funnel O.D. 50 mm
Synonym(s):
Funnels, Standard ground joint funnels
Select a Size
10 EA
R$714.00
R$714.00
Check Cart for Availability
Select a Size
Change View
10 EA
R$714.00
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
R$714.00
Check Cart for Availability
material
polypropylene
Joint
joint ST/NS19/26
packaging
pack of 10 ea
manufacturer/tradename
BRAND 94910
funnel O.D.
50 mm
Related Categories
General description
Half-round body. Stem fits standard ground sockets. Ideal for filling liquids or powders into multi-neck flasks.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)