Documents
BR704851
BRAND® TRANSFERPETTE® S starter kits
(0.5-10ul, 10-100ul, 20-200ul, 100-1000ul, 0.5-5ml), 4 boxes of pipette tips (0.5-20 ul, 2-200 ul, 50-1000 ul, 0.5-5 ml), 1 bench-top rack for 6 TRANSFERPETTE® S
Synonym(s):
Air displacement pipettes, Air interface pipettes, Microliter pipettes, Multichannel pipettes, Pipettes, Piston-operated pipettes, Single channel pipettes
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
sterility
non-sterile
feature
(0.5-10ul, 10-100ul, 20-200ul, 100-1000ul, 0.5-5ml), 4 boxes of pipette tips (0.5-20 ul, 2-200 ul, 50-1000 ul, 0.5-5 ml), 1 bench-top rack for 6 TRANSFERPETTE® S
manufacturer/tradename
BRAND
General description
The Transferpette S microliter pipettes are manual single and multi-channel pipettes for productive and precise pipetting in the laboratory. They offer high quality and many features that make work easier and more efficient, for example: Genuine one-hand operation, short stroke, low operating forces, volume adjustment protection and an easy-to-read volume display. They are easy to maintain and can be adjusted without tools using Easy Calibration technology. The single-channel Transferpette S pipettes cover the volume range from 0.1 ul to 10 ml. The Transferpette S -8 and -12 multichannel pipettes are available in volumes from 0.5 ul to 300 ul.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG
Visit the Document Library
Protocols
This video explains how to adjust the volume on the Transferpette before being ready to pipette.