BR701364
BRAND® sealing film for microplates
self-adhesive, gas permeable, suitable for (for cell and tissue culture)
Synonym(s):
Accessories, PCR, Plates, Sealing films
Select a Size
R$1,478.00
Select a Size
About This Item
R$1,478.00
Recommended Products
material
Rayon
Rayon film
sterility
non-sterile
feature
gas permeable
self-adhesive
packaging
pack of 100 ea
manufacturer/tradename
BRAND 701364
suitability
suitable for (for cell and tissue culture)
Related Categories
General description
Features and Benefits
- Air permeable for optimum oxygen supply
- Secure seal against contamination
- Pierceability for easy sample extraction
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)