Skip to Content
Merck
All Photos(1)

Key Documents

BR701350

BRAND® 96-well deep well plate, stackable

volume 1.2 mL, polypropylene

Synonym(s):

Deep-well Plates, Sample Storage

Sign Into View Organizational & Contract Pricing

Select a Size

24 EA
R$1,602.00

R$1,602.00


In StockDetails



Select a Size

Change View
24 EA
R$1,602.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
material:
polypropylene
round bottom wells
size:
96 well
volume:
1.2 mL
working volume:
1.1 mL
sterility:
non-sterile
binding type:
non-treated surface
feature:
skirt

R$1,602.00


In StockDetails


material

polypropylene
round bottom wells

description

alphanumeric identification system

sterility

non-sterile

feature

skirt

packaging

pack of 42 ea

manufacturer/tradename

BRAND 701350

size

96 well

volume

1.2 mL

working volume

1.1 mL

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • BRAND® 96-well deep well plates are made of high purity polypropylene (PP) which offers excellent chemical resistance.
  • The ′U′ bottom ensures optimal sample mixture and collection.
  • Designed for a range of applications including High-Throughput Screening (HTS), serial dilutions, reagent transfer, and sample storage down to -80 °C.
  • Stackable for easy storage.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service