Key Documents
BR6783
BRAND® replacement amber inspection windows for Titrette®
Synonym(s):
Accessories, Bottle-top Burette, Burettes, Inspection window, Titrette®
Select a Size
2 SETS
R$232.00
R$232.00
Check Cart for Availability
Select a Size
Change View
2 SETS
R$232.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.43
R$232.00
Check Cart for Availability
packaging
pack of 2 SETS
manufacturer/tradename
BRAND 6783
Related Categories
General description
For protection of light-sensitive media, you can replace the clear inspection windows with the amber colored windows (included).
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Titrette is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)