Key Documents
BR65243
BRAND® desiccator ′Novus′
nominal diameter × O.D. × H 250 mm × 320 mm × 425 mm, top socket joint: 24/29
About This Item
UNSPSC Code:
41104510
NACRES:
NB.43
Pricing and availability is not currently available.
Joint
socket joint: top24/29
feature
lid with socket and stopcock
manufacturer/tradename
BRAND 65243
nominal diameter × O.D. × H
250 mm × 320 mm × 425 mm
Related Categories
General description
Boro 3.3. DIN 12491. Precisely ground flat flanges. Base ring surface studded to improve stability and help prevent cracks. Interchangeable lid. Desiccators with sockets are supplied with 24/29 socket, and interchangeable Boro 3.3 stopcock.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)