BR474743
BRAND® microscope slide
ground edges
Synonym(s):
Microscope slides, Microscopy
Select a Size
R$3,133.00
Select a Size
About This Item
R$3,133.00
Recommended Products
material
soda-lime glass
feature
ground edges
manufacturer/tradename
BRAND 474743
size
75 mm × 25 mm × 1 mm
General description
- Made from clear soda-lime glass, hydrolytic class 3.
- Ground edges are recommended to reduce the risk of injury.
- Thickness approx.: 1 mm
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)