BR4600360
BRAND® Dispensette® S Digital bottle-top dispenser
volume range 5-50 mL, without recirculation valve
Synonym(s):
bottle top dispensers
Select a Size
R$7,644.00
Select a Size
About This Item
R$7,644.00
Recommended Products
feature
incremental 0.2 mL
DE-M marking
without recirculation valve
manufacturer/tradename
BRAND 4600360
volume range
5-50 mL
fitting
(GL 32-33)
GL 38
S 40
Related Categories
General description
Features and Benefits
- Extremely easy filling and dispensing
- Simple priming by optimized flow channels
- Rapid and safe volume adjustment with internal scalloped track
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Customers Also Viewed
Protocols
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
Related Content
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.