BR148040
BRAND® PP powder funnel short wide stem
top diam. 150 mm, bottom diam. 35 mm
Synonym(s):
Funnels, Powder funnels
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
BRAND 148040
bottom diam.
35 mm
top diam.
150 mm
Looking for similar products? Visit Product Comparison Guide
Features and Benefits
- BRAND® powder funnel is ideal for transfering powders.
- Made from break-resistant polypropylene, these general lab use funnels include a short wide stem that facilitates optimal filtration.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service