Key Documents
BMSV0020F1
Sigma-Aldrich
Aspire Laboratory Aspirator Accessories
Hydrophobic filter
Select a Size
1 EA
R$247.00
R$247.00
Check Cart for Availability
Select a Size
Change View
1 EA
R$247.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.79
R$247.00
Check Cart for Availability
packaging
pk of 1 ea (1 filter each included)
manufacturer/tradename
Benchmark Scientific V0020-F1
General description
Accessories designed for use with Aspire laboratory aspirator.
Aspire Laboratory Aspirator Accessories, Hydrophobic filter blocks the entering of fluids into the pump unit. It also protects the pump unit from bacterial contamination.
Features and Benefits
  • Provides a barrier to protect against bacterial contamination of pump unit
Legal Information
6538 is a trademark of American Type Culture Collection
Aspire is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)