Skip to Content
Merck
All Photos(1)

Documents

BMSE4000

Benchmark SmartBlue Transilluminator

AC/DC input 115 V

Synonym(s):

SYBR, SmartGlow, blue light, gel imaging, gel casting, transillumination

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106000
NACRES:
NB.13

manufacturer/tradename

Benchmark Scientific E4000

Looking for similar products? Visit Product Comparison Guide

General description

Accuris SmartBlue Transilluminators have been developed for viewing DNA gels stained with new, safe fluorescent dyes. These new dyes, such as SmartGlow, SYBR Green, Gel Green, have an excitation wavelength of 465470nm (visible blue light), and are safer to use than Ethidium Bromide.

Application

Electrophoresis

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service