Skip to Content
Merck
All Photos(1)

Documents

BMSD1133G

Benchmark Bulk Garnet Shards

acid washed

Synonym(s):

grinding medium, grinding beads, beads for homogenization

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13

material

(Garnet shards)

packaging

pkg of 250 g

manufacturer/tradename

Benchmark Scientific D1133-G

compatibility

BeadBug and BeadBlaster

General description

Loose, bulk garnet shards, acid washed for homogenization

Application

bead, homogenization, lysis, garnet, shard, TriplePure, BeadBug, BeadBlaster

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service