Key Documents
BAH500250009
SP Bel-Art® Glove Box Iris Port Entry Cover
includes foam gasket material to create tight seal
Synonym(s):
glove box
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art H50025-0009
General description
Iris port cover works with H50025-0020 Glove Box Iris Port Entry.
Features and Benefits
  • Allows iris ports to be closed off when glove box is not in use
  • Includes foam gasket material to create tight seal
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)