Merck
All Photos(7)
Key Documents
BAF371220020
SP Bel-Art® Spinbar® magnetic stir bar
polygon white PTFE-coated, L × diam. 0.75 in. (20 mm) × 0.25 in. (6 mm)
Synonym(s):
Spinbar® magnetic stir bar
About This Item
UNSPSC Code:
41122800
NACRES:
NB.51
material
polygon white PTFE-coated
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F37122-0020
L × diam.
0.75 in. (20 mm) × 0.25 in. (6 mm)
General description
  • Spinbar® magnetic stir bar is used in conjunction with a magnetic stirring plate for uniform, consistent, and thorough mixing of chemical solutions.
  • Alnico magnet encapsulated in durable and inert FDA grade polytetrafluoroethylene (PTFE) low friction coating.
  • Multi-faceted-shaped bars provide a greater surface area and added turbulence for more vigorous, thorough mixing.
  • Autoclave safe.
Leakproof, seamless, polished, and contains a permanent Alnico V magnet. Egg-shape bar is designed especially for use in round-bottom vessels.
These stirbars have polygon multifaceted surfaces that add turbulence relative to similar smooth size cylindrical bars.
Features and Benefits
  • Center molded pivot ring minimizes the contact area between the bar and the vessel; reduces friction and chattering
  • Marring of plastic containers is diminished
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE coating – Superior Chemical Compatibility
  • Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
  • Autoclave safe
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Spinbar is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)