BAF369110150
SP Bel-Art® Sterileware™ Flat Bottom Sampling Scoop
white polystyrene, nominal capacity 150 mL (5.1 oz), sterile, pack of 100 ea (individually wrapped)
Synonym(s):
Lab Scoop, spoon
About This Item
material
white polystyrene
sterility
sterile
packaging
pack of 100 ea (individually wrapped)
manufacturer/tradename
Bel-Art F36911-0150
bowl H × L × W
1.375 in. (34 mm) × 4.875 in. (123 mm) × 2.75 in. (68 mm)
handle L
3.75 in. (9.5 cm)
nominal capacity
150 mL (5.1 oz)
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Handle bump acts as thumb rest to help keep the hand away from the scoop; reduces contamination risks
- Flat bottom allows for use as a weigh boat
- Approximate volumes can be measured by removing excess powder
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- High impact polystyrene (HIPS) scoops heat sealed in individual polyethylene bags
- Gamma irradiated sterile (SAL 10^-6)
- Ready for one time use and disposal
- Lot stamped for accurate tracing
Note: Sterilized products are not returnable.
These sampling scoops are designed for retrieving samples from a larger source. Capacity/volumes are estimated and are not meant for taking exact sample weights/volumes.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service