BAF132040001
Cover for glass disposal carton
autoclavable
Select a Size
R$371.00
Select a Size
About This Item
R$371.00
Recommended Products
feature
autoclavable
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F13204-0001
General description
- Opens with a Simple Touch
- Sanitary and reusable
- Green center panel matches printing on glass disposal cartons providing fast visual recognition of the waste stream
- Features smooth 10.1 x 10.1 cm (4 x 4 in.) area for applying labels
- Provides a wider opening 20.3 x 24.1 cm (8 in. x 9 1/2 in.) permitting safer and more sanitary deposit of wastes into the container
Choose from one of the most recent versions:
Certificates of Analysis (COA)