Skip to Content
Merck
All Photos(1)

Documents

B65403

Sigma-Aldrich

Bromodiphenylmethane

95%

Synonym(s):

Benzhydryl bromide, Diphenylbromomethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2CHBr
CAS Number:
Molecular Weight:
247.13
Beilstein:
638544
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

bp

184 °C/20 mmHg (lit.)

mp

35-39 °C (lit.)

SMILES string

BrC(c1ccccc1)c2ccccc2

InChI

1S/C13H11Br/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10,13H

InChI key

OQROAIRCEOBYJA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service