Skip to Content
Merck
All Photos(3)

Key Documents

B34702

Sigma-Aldrich

Biphenyl-2-carboxylic acid

98%

Synonym(s):

2-Phenylbenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4CO2H
CAS Number:
Molecular Weight:
198.22
Beilstein:
974075
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

bp

199 °C/10 mmHg (lit.)

mp

111-113 °C (lit.)

SMILES string

OC(=O)c1ccccc1-c2ccccc2

InChI

1S/C13H10O2/c14-13(15)12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-9H,(H,14,15)

InChI key

ILYSAKHOYBPSPC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Tiffany R Walsh et al.
Chemical reviews, 117(20), 12641-12704 (2017-08-30)
Peptide sequences are known to recognize and bind different nanomaterial surfaces, which has resulted in the screening and identification of hundreds of peptides with the ability to bind to a wide range of metallic, metal oxide, mineral, and polymer substrates.
Heleen M Kool et al.
American journal of physiology. Lung cellular and molecular physiology, 317(3), L317-L331 (2019-07-04)
The mortality and morbidity of patients with congenital diaphragmatic hernia (CDH) is primarily caused by treatment-resistant, persistent pulmonary hypertension. Structural vascular changes, exemplified by extensive muscularization, are already present early in gestation, but the origin of these abnormalities is unknown.
A N Baeza et al.
Journal of chromatography. A, 1474, 121-129 (2016-11-07)
This work reports the preparation of molecularly imprinted polymers (MIPs) selective to cephalosporin (CF) antibiotics, and their application as molecularly imprinted solid-phase extraction (MISPE) sorbents for the determination of these antimicrobials in milk samples. Several functional monomers and cross-linkers have
Javier L Urraca et al.
Analytical and bioanalytical chemistry, 408(7), 1843-1854 (2015-09-08)
This work describes the development and application of class-selective molecularly imprinted polymers (MIPs) for the analysis of beta-lactamase-resistant penicillins, namely cloxacillin (CLOXA), oxacillin (OXA), and dicloxacillin (DICLOXA), in milk samples. Our method is based on molecularly imprinted solid-phase extraction (MISPE)

Questions

  1. What is the use of this molecule in pharma industry

    1 answer
    1. Product B34702 is provided for research and development use. There are numerous uses for this compound in research applications. Please see a couple of citations below that you may find beneficial.

      Oxidative Ring-Opening of Aromatics: Decomposition of Biphenyl Carboxylic Acids and Zinc Biphenyl Carboxylates
      Natalia Montoya Sánchez and Arno de Klerk
      Energy & Fuels 2015 29 (12), 7910-7922
      DOI: 10.1021/acs.energyfuels.5b02066

      Harnessing the Energy of Visible Light to Promote LMCT of Cu(II) Species for Intramolecular Dehydrogenative Lactonization of Biaryl-2-carboxylic Acids
      Partha Pratim Sen and Sudipta Raha Roy
      Organometallics 2023 42 (13), 1658-1666
      DOI: 10.1021/acs.organomet.3c00210

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service