Skip to Content
Merck
All Photos(1)

Key Documents

ALD01784

Sigma-Aldrich

N-(tert-butyl)-2-methoxypyridin-3-amine

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
R$1,092.00

R$1,092.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
R$1,092.00

About This Item

MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

R$1,092.00


Check Cart for Availability

Request a Bulk Order

form

liquid

storage temp.

2-8°C

SMILES string

CC(C)(C)NC1=CC=CN=C1OC

InChI

1S/C10H16N2O/c1-10(2,3)12-8-6-5-7-11-9(8)13-4/h5-7,12H,1-4H3

InChI key

KNFHUHWGLSSVFV-UHFFFAOYSA-N

Application

Traditionally challenging to access, this hindered amine provides a chemically differentiated building block for organic synthesis and medicinal chemistry, specifically for preparation of drug candidates containing hindered amine motifs. Additionally, this and over 80 other sterically hindered secondary amines in the Sigma-Aldrich portfolio provide structurally distinct screening compounds. Screen several through Aldrich Market Select. This hindered amine was accessed via a recent publication reported by Baran and coworkers using an innovative hydroamination method.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Baran Group offers zinc-based reagents for versatile transformations in organic synthesis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service