Skip to Content
Merck
All Photos(1)

Key Documents

ALD00034

Sigma-Aldrich

(S)-tert-Butyl 1-(chloromethyl)-5-hydroxy-1H-benzo[e]indole-3(2H)-carboxylate

95% (HPLC)

Synonym(s):

N-Boc-seco-CBI

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
R$500.00

R$500.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
R$500.00

About This Item

Empirical Formula (Hill Notation):
C18H20ClNO3
CAS Number:
Molecular Weight:
333.81
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

R$500.00


Check Cart for Availability

Request a Bulk Order

Assay

95% (HPLC)

form

powder

optical activity

[α]/D -89.0, c = 0.05% in chloroform

storage temp.

−20°C

SMILES string

OC1=C2C(C=CC=C2)=C3C(N(C(OC(C)(C)C)=O)C[C@H]3CCl)=C1

InChI

1S/C18H20ClNO3/c1-18(2,3)23-17(22)20-10-11(9-19)16-13-7-5-4-6-12(13)15(21)8-14(16)20/h4-8,11,21H,9-10H2,1-3H3/t11-/m1/s1

InChI key

DGPQGWRHSMFTSW-LLVKDONJSA-N

Application

N-Boc-CBI best used as its seco precursor (seco-N-Boc-CBI) is a simple alkylation subunit analog of that found in the naturally occurring antitumor agents the duocarmycins, yatakemycin, and CC-1065, which act by sequence selective DNA alkylation.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Boger Lab utilizes nature's solutions in complex natural products, extending them through rational design for improved biological outcomes.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service