Skip to Content
Merck
All Photos(2)

Key Documents

A8201

Sigma-Aldrich

4-Acetamidothiophenol

≥90%

Synonym(s):

4′-Mercaptoacetanilide

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$351.00

R$351.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
R$351.00

About This Item

Linear Formula:
CH3CONHC6H4SH
CAS Number:
Molecular Weight:
167.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$351.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥90%

form

powder

mp

150-153 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=O)Nc1ccc(S)cc1

InChI

1S/C8H9NOS/c1-6(10)9-7-2-4-8(11)5-3-7/h2-5,11H,1H3,(H,9,10)

InChI key

AYEQJLOHMLYKAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandro Mosco et al.
PloS one, 7(1), e30052-e30052 (2012-01-19)
The crustacean Hyperglycemic Hormone (cHH) is present in many decapods in different isoforms, whose specific biological functions are still poorly understood. Here we report on the first chemical synthesis of three distinct isoforms of the cHH of Astacus leptodactylus carried
Cristiano Matricardi et al.
ACS nano, 12(8), 8531-8539 (2018-08-15)
Metal colloids are of great interest in the field of nanophotonics, mainly due to their morphology-dependent optical properties, but also because they are high-quality building blocks for complex plasmonic architectures. Close-packed colloidal supercrystals not only serve for investigating the rich
Ivan V Smolyaninov et al.
Bioorganic chemistry, 89, 103003-103003 (2019-05-28)
A number of asymmetrical thioethers based on 3,5-di-tert-butylcatechol containing sulfur atom bonding with physiologically active groups in the sixth position of aromatic ring have been synthesized and the electrochemical properties, antioxidant, cryoprotective activities of new thioethers have been evaluated. Cyclic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service