Skip to Content
Merck
All Photos(2)

Key Documents

A74602

Sigma-Aldrich

3-Aminophenyl sulfone

97%

Synonym(s):

3,3′-Sulfonyldianiline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2NC6H4)2SO2
CAS Number:
Molecular Weight:
248.30
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

powder

mp

170-173 °C (lit.)

SMILES string

Nc1cccc(c1)S(=O)(=O)c2cccc(N)c2

InChI

1S/C12H12N2O2S/c13-9-3-1-5-11(7-9)17(15,16)12-6-2-4-10(14)8-12/h1-8H,13-14H2

InChI key

LJGHYPLBDBRCRZ-UHFFFAOYSA-N

General description

3-Aminophenyl sulfone is an amine based curing agent that facilitates the formation of heat resistant epoxy based adhesive system. It has an amine hydrogen equivalent weight of 62.[1][2]

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - STOT RE 2

Target Organs

Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of the structure of the aromatic curing agent on the cure kinetics of epoxy networks.
Grillet AC, et al.
Polymer, 30(11), 2094-2103 (1989)
Studies on the adhesive properties of solid elastomer-modified novolac epoxy resin.
Gouri C, et al.
International Journal of Adhesion and Adhesives, 20(4), 305-314 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service