Skip to Content
Merck
All Photos(1)

Documents

94959

Sigma-Aldrich

Vinylcyclooctane

≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18
CAS Number:
Molecular Weight:
138.25
Beilstein:
4364365
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0%

form

liquid

SMILES string

C=CC1CCCCCCC1

InChI

1S/C10H18/c1-2-10-8-6-4-3-5-7-9-10/h2,10H,1,3-9H2

InChI key

UPHVHMSLLBDZEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

69.8 °F - closed cup

Flash Point(C)

21 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P G Gervasi et al.
Toxicology letters, 16(3-4), 217-223 (1983-05-01)
Vinylcyclooctane, when administered to mice at 500 mg/kg, produced reduction of microsomal cytochrome P-450, heme, aminopyrine-N-demethylase and ethoxycoumarin-O-deethylase activities with respect to control values; furthermore the hepatic reduced glutathione level was depleted suggesting that glutathione is involved in the vinylcyclooctane
P G Gervasi et al.
Toxicology letters, 20(3), 243-249 (1984-03-01)
Vinylcyclooctane (VCO), which binds to the active site of cytochrome P-450 (P-450) giving a type I difference spectrum, has been found to form the corresponding epoxide as the main metabolite on treatment with liver microsomal monooxygenase obtained from phenobarbital-treated or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service