Skip to Content
Merck
All Photos(1)

Key Documents

94866

Sigma-Aldrich

Boc-α-(4-bromobenzyl)-DL-Pro-OH

≥96.0% (HPLC)

Synonym(s):

1-Boc-2-(4-bromobenzyl)-2-pyrrolidinecarboxylic acid, Boc-α-(4-bromobenzyl)-DL-proline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22BrNO4
CAS Number:
Molecular Weight:
384.26
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

product name

Boc-α-(4-bromobenzyl)-DL-Pro-OH, ≥96.0% (HPLC)

Assay

≥96.0% (HPLC)

form

powder

color

off-white

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N1CCCC1(Cc2ccc(Br)cc2)C(O)=O

InChI

1S/C17H22BrNO4/c1-16(2,3)23-15(22)19-10-4-9-17(19,14(20)21)11-12-5-7-13(18)8-6-12/h5-8H,4,9-11H2,1-3H3,(H,20,21)

InChI key

VQWRXYZZPMLJRL-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service