Key Documents
930318
Sigma-Aldrich
TEM Grids
Formvar Supported Copper Square Mesh, grid size 200 mesh, box of 25
Synonym(s):
TEM grids
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Pricing and availability is not currently available.
Product Name
Formvar Supported Copper Square Mesh, grid size 200 mesh, box of 25
material
copper grid
formvar film
description
film layer thickness 10 nm
packaging
box of 25
grid size
200 mesh
General description
A 10 nm thin formvar film supported standard copper square mesh grids. Copper grid, 200 mesh. All of our film is laid on the shiny side of the grid.
Choose from one of the most recent versions:
Certificates of Analysis (COA)