Skip to Content
Merck
All Photos(1)

Documents

929581

Sigma-Aldrich

PCL-PEG-PCL diacrylate

new

average Mn 2000-20000-2000, poly(caprolactone)-b-poly(ethylene glycol)-b-poly(caprolactone) diacrylate

Synonym(s):

Acrylate-PCL-PEG-PCL-acrylate, Biodegradable triblock copolymer, PCL-b-PEG-b-PCL, PEG hydrogel, PEGDA

Sign Into View Organizational & Contract Pricing


About This Item

Quality Level

form

low-melting solid

mol wt

PCL Mn 2000
PEG Mn 20000
24000 (theoretical)
average Mn 2000-20000-2000

contains

≤300 ppm MEHQ as inhibitor

color

white to off-white

Ω-end

acrylate

α-end

acrylate

storage temp.

−20°C

SMILES string

O=C(OCCOC(CCCCCOC(C=C)=O)=O)CCCCCOC(C=C)=O

Looking for similar products? Visit Product Comparison Guide

General description

PCL-PEG-PCL diacrylate, average Mn 2000-20000-2000, is a functionalized, amphiphilic triblock copolymer composed of a central hydrophilic, biocompatible poly(ethylene glycol) (PEG) block and two terminal hydrophobic, biodegradable poly(caprolactone) (PCL) blocks. PCL-PEG-PCL diacrylate can be crosslinked via free radical polymerization and/or photopolymerication to form a crosslinked hydrogel network. Hydrolysis of the esters of the PCL blocks permits degradation of the crosslinked networks.

Application

  • Tissue engineering
  • 3D printing
  • Drug delivery

Features and Benefits

  • PCL-PEG-PCL triblock copolymer
  • Hydrolytically degradable PCL blocks
  • Acrylate functional groups permit photopolymerization

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service