Skip to Content
Merck
All Photos(1)

Key Documents

929115

Sigma-Aldrich

Maleimide-PEG-Propionic Acid

~ 4 PEG

Synonym(s):

Discrete PEG, Mal PEG4 acid, Monodisperse PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H3O2(C6H10OO)n(C2H4O)m(OC6H10O)nOC2H3
MDL number:
UNSPSC Code:
12352106

Quality Level

form

viscous liquid

mol wt

estimated mol wt ~416.4 Da

color

colorless to light yellow

Ω-end

maleimide (Mal)

α-end

(propionic acid (PA))

storage temp.

−20°C

General description

Maleimide PEG4 Propionic Acid, MW 416.4 (Mal-PEG4-PA) is a heterobifunctional monodisperse poly(ethylene glycol) (PEG) crosslinking agent or spacer. The maleimide functional group reacts with sulfhydryl groups via Michael addition reaction. The propionic acid functional group reacts with free amines to form amide bonds.
The Peg product has a low melting point. The physical appearance of the product may change with the envrionmental temperature. The change in physical appearance will not affect the use of the product.

Application

  • Improves water solubility
  • Prolongs the half-life and reduces drug immunogenicity and toxicity in drug delivery applications
  • PEGylation and conjugation of proteins, peptides, and antibodies
  • PEG-modified functional coatings of nanoparticles or microparticles

Features and Benefits

  • PEG crosslinker that joins sulfhydryl groups to free amines through a monodisperse PEG of specific molecular weight and number of repeat units
  • Maleimide (mal) reacts with sulfhydryl groups, thiol, and mercapto via Michael addition reaction
  • Propionic acid (PA) reacts with free amines using EDC or other carbodiimides to form amide bonds
  • Monodisperse/discrete PEGs lack the polydispersity of traditional PEG polymers
  • Enables the conjugation of two molecules in aqueous solution
  • Excellent water solubility, fast kinetics, and stability in aqueous buffers
  • Biocompatible

Legal Information

Product of JenKem Technology

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service