Skip to Content
Merck
All Photos(3)

Key Documents

926299

Sigma-Aldrich

Micro Vortex Mixer Chip

Fluidic 642, PC

Synonym(s):

Microfluidic chip

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
R$849.00

R$849.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
R$849.00

About This Item

UNSPSC Code:
42142600
NACRES:
NA.23

R$849.00


Check Cart for Availability

Request a Bulk Order

description

Microfludic chip x1

Looking for similar products? Visit Product Comparison Guide

Application

The Vortex Mixer Fluidic 642 PC microfluidic chip applies passive mixing principles and sheath fluidic flow to ensure mixing. Vortex Mixer Fluidic 642 is one of three vortex mixer chip designs.

Features and Benefits

  • Ten identical micromixing units per chip
  • Each unit features one outlet, a central inlet for solution A and another inlet for solution B
  • Solutions A and B mix and split into two lateral fluidic streams via sheath flow
  • To improve mixing, daisy-chaining of serval vortex units is recommended

Components

Chip Properties:
  • Mini Luer Interface
  • Material: Polycarbonate (PC)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongfei Liu et al.
Lab on a chip, 17(11), 1856-1883 (2017-05-10)
The microfluidic technique has brought unique opportunities toward the full control over the production processes for drug delivery carriers, owing to the miniaturisation of the fluidic environment. In comparison to the conventional batch methods, the microfluidic setup provides a range
Sharma T Sanjay et al.
Advanced drug delivery reviews, 128, 3-28 (2017-09-19)
Conventional systematically-administered drugs distribute evenly throughout the body, get degraded and excreted rapidly while crossing many biological barriers, leaving minimum amounts of the drugs at pathological sites. Controlled drug delivery aims to deliver drugs to the target sites at desired

Articles

Microfluidic assembly improves polyamine nanoencapsulation of nucleic acids, overcoming challenges like polydispersity and poor reproducibility.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service