Skip to Content
Merck
All Photos(1)

Key Documents

924172

Sigma-Aldrich

Tris[(1,1,1,3,3,3-hexafluoropropan-2-yl)oxy]silane

Synonym(s):

HSi(OCH(CF3)2)3

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
R$1,114.00
1 G
R$2,749.00

R$1,114.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
R$1,114.00
1 G
R$2,749.00

About This Item

Empirical Formula (Hill Notation):
C9H4F18O3Si
CAS Number:
Molecular Weight:
530.18
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

R$1,114.00


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

refractive index

n/D 1.284

density

1.609 g/mL

functional group

fluoro

Related Categories

Application

Tris[(1,1,1,3,3,3-hexafluoropropan-2-yl)oxy]silane is a silylating reagent used to silylate the C-terminus of unprotected amino acids and peptides, allowing for coupling with N-silylated amino acids and peptides to form discrete peptide bonds using a one-pot method.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wataru Muramatsu et al.
Journal of the American Chemical Society, 143(18), 6792-6797 (2021-05-01)
A one-pot peptide bond-forming reaction has been developed using unprotected amino acids and peptides. Two different silylating reagents, HSi[OCH(CF3)2]3 and MTBSTFA, are instrumental for the successful implementation of this approach, being used for the activation and transient masking of unprotected

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service