924113
(R,E)-1-(2-Aminoethyl)cyclooct-4-en-1-ol, 5-ax-Hydroxy-5-eq-(2-aminoethyl)-trans-cyclooctene
![New Product new](/static/new_product_badge.png)
Synonym(s):
Amine-a-TCO
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H19NO
Molecular Weight:
169.26
storage temp.
−20°C
Quality Level
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jessica E Pigga et al.
Angewandte Chemie (International ed. in English), 60(27), 14975-14980 (2021-03-21)
trans-Cyclooctenes (TCOs) are essential partners in the fastest known bioorthogonal reactions, but current synthetic methods are limited by poor diastereoselectivity. Especially hard to access are hydrophilic TCOs with favorable physicochemical properties for live cell or in vivo experiments. Described is a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service