Skip to Content
Merck
All Photos(1)

Key Documents

922633

Sigma-Aldrich

5-Methyl-2-(3-thienyl)pyridine

≥95%

Synonym(s):

Chattopadhyay′s PYT Ligand

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
R$868.00
500 MG
R$2,715.00

R$868.00


0 left in stock

Request a Bulk Order

Select a Size

Change View
100 MG
R$868.00
500 MG
R$2,715.00

About This Item

Empirical Formula (Hill Notation):
C10H9NS
CAS Number:
Molecular Weight:
175.25
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

R$868.00


0 left in stock

Request a Bulk Order

Quality Level

Assay

≥95%

form

powder

mp

45-47 °C

storage temp.

−20°C

SMILES string

[s]1cc(cc1)c2ncc(cc2)C

InChI

1S/C10H9NS/c1-8-2-3-10(11-6-8)9-4-5-12-7-9/h2-7H,1H3

InChI key

UXJGBSHKYRDOPU-UHFFFAOYSA-N

Application

5-Methyl-2-(3-thienyl)pyridine is a ligand for the Ir-catalyzed directed C(sp2)-H and C(sp3)-H borylation of a diverse set of aryl, heteroaryl, and alkyl substrates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Md Emdadul Hoque et al.
Journal of the American Chemical Society, 143(13), 5022-5037 (2021-03-31)
Here we describe the discovery of a new class of C-H borylation catalysts and their use for regioselective C-H borylation of aromatic, heteroaromatic, and aliphatic systems. The new catalysts have Ir-C(thienyl) or Ir-C(furyl) anionic ligands instead of the diamine-type neutral

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service