Skip to Content
Merck
All Photos(1)

Key Documents

918830

SAFC

Sodium cyanoborohydride

Synonym(s):

Sodium cyanotrihydridoborate

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
R$714.00
100 G
R$5,147.00
1 KG
R$45,049.00

R$714.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
R$714.00
100 G
R$5,147.00
1 KG
R$45,049.00

About This Item

Linear Formula:
NaBH3CN
CAS Number:
Molecular Weight:
62.84
Beilstein:
4152656
MDL number:
UNSPSC Code:
12352306

R$714.00


Check Cart for Availability

Request a Bulk Order

biological source

synthetic

Quality Level

mp

>242 °C (dec.) (lit.)

suitability

suitable for manufacturing use

SMILES string

[Na+].[BH3-]C#N

InChI

1S/CH3BN.Na/c2-1-3;/h2H3;/q-1;+1

InChI key

CVDUGUOQTVTBJH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Our comprehensive portfolio of downstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Ranging from non-GMP grades for low-risk application, to IPEC-PQG GMP for higher-risk applications, we have products covering all your manufacturing needs.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Sol. 1 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service