Skip to Content
Merck
All Photos(1)

Key Documents

914053

Sigma-Aldrich

Lead(II) chloride

AnhydroBeads, 99.999%, (perovskite grade)

Synonym(s):

Cotunnite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PbCl2
CAS Number:
Molecular Weight:
278.11
MDL number:
UNSPSC Code:
12352302
Pricing and availability is not currently available.

vapor pressure

1 mmHg ( 547 °C)

Quality Level

product line

AnhydroBeads

Assay

99.999%

form

powder

particle size

-10 mesh

bp

950 °C (lit.)

mp

501 °C (lit.)

density

5.85 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

Cl[PbH2]Cl

InChI

1S/2ClH.Pb/h2*1H;/q;;+2/p-2

InChI key

HWSZZLVAJGOAAY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Lead chloride finds application in synthesis of perovksites based photovoltaic materials. Our perovskite grade PbBr2 can readily be dissolved in DMF: DMSO (1:1) to yield 0.4 M solution.
Lead(II) Chloride is a crucial precursor material for the synthesis of the perovskite absorber layer in perovskite solar cells. It plays a vital role in the absorption of light in the perovskite absorber layer.

Packaging

Packed in ampules

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Not finding the right product?  

Try our Product Selector Tool.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 1A - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service