Skip to Content
Merck
All Photos(3)

Documents

913472

Sigma-Aldrich

(2S,2S′,3S,3S′)-WingPhos

≥95%

Synonym(s):

(2S,2′S,3S,3′S)-4,4′-Di(anthracen-9-yl)-3,3′-di-tert-butyl-2,2′,3,3′-tetrahydro-2,2′-bibenzo[d][1,3]oxaphosphole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H44O2P2
CAS Number:
Molecular Weight:
738.83
UNSPSC Code:
12352200

Assay

≥95%

form

powder or crystals

optical purity

ee: ≥99% (HPLC)

reaction suitability

reagent type: ligand

functional group

phosphine

Application

(2S,2S′,3S,3S′)-WingPhos is a P-chiral biphosphorus ligand for Rh-catalyzed asymmetric hydrogenations as well as Rh-catalyzed asymmetric addition of aryl boroxines to ketones.

Product can be used with our benchtop hydrogen generator, H-Genie Lite (Z744083)

Legal Information

Sold in collaboration with Zejun Pharmaceuticals

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design of phosphorus ligands with deep chiral pockets: practical synthesis of chiral β-arylamines by asymmetric hydrogenation.
Guodu Liu et al.
Angewandte Chemie (International ed. in English), 52(15), 4235-4238 (2013-03-16)
Linwei Huang et al.
Angewandte Chemie (International ed. in English), 55(14), 4527-4531 (2016-03-05)
Highly enantioselective additions of arylboroxines to simple aryl ketones have been achieved for the first time with a Rh/(R,R,R,R)-WingPhos catalyst, thus providing a range of chiral diaryl alkyl carbinols with excellent ee values and yields. (R,R,R,R)-WingPhos has been proven to be
Jinbin Zhu et al.
Angewandte Chemie (International ed. in English), 58(45), 16119-16123 (2019-08-31)
Highly enantioselective rhodium-catalyzed addition of arylboroxines to N-unprotected ketimines is realized for the first time by employing chiral BIBOP-type ligands with a Rh loading as low as 1 mol %. A range of chiral α-trifluoromethyl-α,α-diaryl α-tertiary amines or 3-amino-3-aryloxindoles were formed with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service