Skip to Content
Merck
All Photos(2)

Key Documents

911143

Sigma-Aldrich

HO-PEG24-propanoic acid t-butyl ester

Synonym(s):

HO-PEG-tert-butyl ester, Hydroxyl PEG tert-butyl ester, Hydroxyl-PEG-tert-butyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
R$2,866.00

R$2,866.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
R$2,866.00

About This Item

Empirical Formula (Hill Notation):
C55H110O27
CAS Number:
Molecular Weight:
1203.45
MDL number:
UNSPSC Code:
12352108
NACRES:
NA.23

R$2,866.00


Check Cart for Availability

Request a Bulk Order

form

solid or liquid

mol wt

average Mn 1,200 (by NMR)

reaction suitability

reagent type: chemical modification reagent
reactivity: amine reactive

color

white to off-white

Ω-end

(tert-Butyl ester)

α-end

hydroxyl

polymer architecture

shape: linear
functionality: monofunctional

storage temp.

−20°C

SMILES string

O(CCO)CCC(=O)OC(C)(C)C

InChI

1S/C9H18O4/c1-9(2,3)13-8(11)4-6-12-7-5-10/h10H,4-7H2,1-3H3

InChI key

ACWLDITVZGWIKA-UHFFFAOYSA-N

Application

Polyethylene glycols (PEGs) are FDA approved hydrophilic polymers. They are non-toxic, non-immunogenic and non-antigenic, and are widely used in a varity of biomedical applications such as bioconjugation, drug delivery, surface functionalization, and tissue engineering. The propionic acid group in HO-PEG-propanoic acid t-butyl ester is protected, preventing self-coupling.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

CRISPR/Cas9 delivery via nonviral nanoparticles shows promising advancements for gene editing in disease treatment.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service