Skip to Content
Merck
All Photos(1)

Key Documents

908665

Sigma-Aldrich

Acryloxyethyl thiocarbamoyl rhodamine B

Synonym(s):

Fluorescent monomer, Rhodamine B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H38N3O6SCl
Molecular Weight:
652.20
UNSPSC Code:
12352116

form

(powder with shiny flecks)

storage condition

protect from light

color

brown

storage temp.

−20°C

Application

Acryloxyethyl thiocarbamoyl rhodamine B is a fluorescent monomer for synthesizing fluorescent polymers. Fluorescent polymers are especially useful in the preparation of fluorescent microspheres or nanoparticles, for bioimaging or drug delivery research.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elution of Labile Fluorescent Dye from Nanoparticles during Biological Use.
Tenuta T, et al.
PLoS ONE (2011)
Fluorescent and chemico-fluorescent responsive polymers from dithiomaleimide and dibromomaleimide functional monomers.
Robin M P et al.
Chemical Science, 5, 2717-2717 (2014)
Fluorescent and chemico-fluorescent responsive polymers from dithiomaleimide and dibromomaleimide functional monomers.
Robin M P et al.
Chemical Science, 5, 2717-2723 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service