Skip to Content
Merck
All Photos(1)

Key Documents

907197

Sigma-Aldrich

1,4-Dinitro-1-imidazole

Synonym(s):

1,4-DNIm, 1,4-Dinitroimidazole, Bifunctional bioconjugation reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H2N4O4
CAS Number:
Molecular Weight:
158.07
MDL number:
UNSPSC Code:
12352005

Assay

≥95%

form

solid

storage temp.

−20°C

InChI

1S/C3H2N4O4/c8-6(9)3-1-5(2-4-3)7(10)11/h1-2H

InChI key

HZPSREFSUPXCMN-UHFFFAOYSA-N

Application

1,4-Dinitro-1-imidazole (1,4-DNIm) and other dinitroimidazoles were demonstrated to be bifunctional bioconjugation reagents for protein functionalization and peptide macrocyclization. In acidic to neutral aqueous conditions, 1,4-DNIms chemoselectively react with cysteines; in organic solvents in the presence of base, 1,4-DNIms chemoselectively react with lysines to form stable conjugates

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service