Skip to Content
Merck
All Photos(1)

Key Documents

905690

Sigma-Aldrich

Dimethylsulfoxonium-2-(chloro)benzoylmethylide

≥95%

Synonym(s):

1-(2-Chlorophenyl)-2-(dimethyl(oxo)-sulfaneylidene)ethan-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11ClO2S
CAS Number:
Molecular Weight:
230.71
UNSPSC Code:
12352101

Assay

≥95%

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

136-137 °C

storage temp.

−20°C

SMILES string

O=C(C1=CC=CC=C1Cl)C=S(C)(C)=O

General description

Sulfoxonium ylides are highly stable carbene precursors, serving as a safer alternative to diazo compounds as they that do not generate gases as byproducts. Sulfoxonium ylides have been shown to be effective in a wide array of transition metal catalyzed C-H functionalization and coupling reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexánder Garay Talero et al.
Organic letters, 20(22), 7206-7211 (2018-10-27)
A general, mild, and versatile synthesis of the challenging α-aryl-β-ketosulfoxonium ylides has been developed for the first time, substituting traditional methods starting from diazo compounds. The arylation of easily accessible β-ketosulfoxonium ylides using aryne chemistry allowed the preparation of a
Youwei Xu et al.
Organic letters, 19(16), 4307-4310 (2017-08-08)
Direct and efficient synthesis of 1-naphthols has been realized via Rh(III)-catalyzed C-H activation of sulfoxonium ylides and subsequent annulation with alkynes, where the sulfoxonium ylide functioned as a new traceless bifunctional directing group. This reaction occurred under redox-neutral conditions with
Panjie Hu et al.
Organic letters, 20(8), 2160-2163 (2018-04-03)
Rh(III)-catalyzed cascade C-H activation of benzoylacetonitriles and annulation with sulfoxonium ylides was realized, leading to selective synthesis of naphthols, 2,3-dihydronaphtho[1,8- bc]pyrans, and naphtho[1,8- bc]pyrans. This step-economic reaction proceeded efficiently under mild and redox-neutral conditions via multiple C-H activations.

Related Content

Ketosulfoxonium ylides serve as versatile carbene equivalents for large-scale reactions, releasing dimethyl sulfoxide biproducts.

Ellman group developed electron-rich phosphine ligands for C-H functionalization and tert-Butanesulfinamide for asymmetric amine synthesis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service