Skip to Content
Merck
All Photos(2)

Key Documents

901904

Sigma-Aldrich

tBuXPhos

95%

Synonym(s):

2-Di-tert-butylphosphino-2′,4′,6′-triisopropylbiphenyl, t-Bu XPhos, tert-Butyl XPhos

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$333.00
5 G
R$1,333.00
25 G
R$3,407.00

R$333.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
R$333.00
5 G
R$1,333.00
25 G
R$3,407.00

About This Item

Empirical Formula (Hill Notation):
C29H45P
CAS Number:
Molecular Weight:
424.64
MDL number:
UNSPSC Code:
12352002
NACRES:
NA.22

R$333.00


Check Cart for Availability

Request a Bulk Order

Assay

95%

form

powder or crystals

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

reagent type: ligand

mp

148-151 °C (lit.)
150 °C

functional group

phosphine

SMILES string

CC(P(C(C=CC=C1)=C1C(C(C(C)C)=CC(C(C)C)=C2)=C2C(C)C)C(C)(C)C)(C)C

InChI

1S/C29H45P/c1-19(2)22-17-24(20(3)4)27(25(18-22)21(5)6)23-15-13-14-16-26(23)30(28(7,8)9)29(10,11)12/h13-21H,1-12H3

InChI key

SACNIGZYDTUHKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

tBuXPhos [2-Di-tert-butylphosphino-2′,4′,6′-triisopropylbiphenyl] is an air-stable, electron-rich biaryl phosphine ligand developed by the Buchwald group to enhance the reactivity of palladium catalysis during cross-coupling reactions.

Application

tBuXPhos has been used in the preparation of [tBuXPhosAu(MeCN)]BAr4F, a gold catalyst for the intermolecular [2+2] cycloaddition of terminal arylalkynes with substituted alkenes to form functionalized cyclobutenes with high regioselectivity.

Legal Information

Usage subject to Patents: EP 1097158; JP 5758844; CA 2336691

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service