Skip to Content
Merck
All Photos(1)

Key Documents

901603

Sigma-Aldrich

Bromoacetic acid solution

1 M in dichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2COOH
CAS Number:
Molecular Weight:
138.95
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

form

liquid

concentration

1 M in dichloromethane

refractive index

n/D 1.4295

density

1.3429 g/mL

SMILES string

BrCC(O)=O

InChI

1S/C2H3BrO2/c3-1-2(4)5/h1H2,(H,4,5)

InChI key

KDPAWGWELVVRCH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Valuable building block offered as a solution in dichloromethane for more convenient handling.

related product

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service