Skip to Content
Merck
All Photos(1)

Key Documents

900722

Sigma-Aldrich

Bovine tendon powder

long fiber, Type I collagen, ≥60% collagen, suitable for biomedical research

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41106305
NACRES:
NA.23

Quality Level

description

Electrophoretic pattern (purity by SDS PAGE): α, β, and γ bands present
Hydroxyproline assay: ≥60% collagen w/w

form

powder

shelf life

24 mo. (after date of manufacture)

impurities

<10 EU/g Endotoxin

color

white to off-white

pH

3.7-5.7

transition temp

Tm 50-70 °C

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Bovine tendon powder is an acidic (pH 3.7 - 5.7) milled type I collagen powder that is ideal for use in custom formulations. This material is nonabsorbent, but highly wetable making it ideal for orthopedic research applications.

Application

Collagen is a major structural protein that provides structure and strength to tissues in the body. Due to its biocompatibility, biodegradability, and high abundance, collagen has been utilized for many applications in biomedical research. Applications of fibrillar collagen may include:
  • Development of collagen-based tissues for skin replacement, bone substitutes, and to generate artificial blood vessels and valves.
  • Drug delivery systems such as sponges for burns/wounds, formulations with liposomes for sustained drug delivery, materials for transdermal drug delivery, and nanoparticles for gene delivery.
  • As collagen scaffolds for research in cancer, electrophysiological protocols, neuron myelination, immunology, and other cell culture applications.

Packaging

1EA=1G

Legal Information

Product of Collagen Solutions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Collagen-based delivery systems effectively deliver a range of viral and non-viral gene carriers by providing spatiotemporal control over delivery and reducing degradation rates and immune responses.

Professor Shrike Zhang (Harvard Medical School, USA) discusses advances in 3D-bioprinted tissue models for in vitro drug testing, reviews bioink selections, and provides application examples of 3D bioprinting in tissue model biofabrication.

Professor Shrike Zhang (Harvard Medical School, USA) discusses advances in 3D-bioprinted tissue models for in vitro drug testing, reviews bioink selections, and provides application examples of 3D bioprinting in tissue model biofabrication.

Protocols

Frequently asked questions (FAQs) for KAPA SYBR® FAST One-Step qRT-PCR Kits.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service