Skip to Content
Merck
All Photos(1)

Key Documents

900408

Sigma-Aldrich

Boron nitride

nanoplatelet, lateral dimensions <5 μ

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BN
CAS Number:
Molecular Weight:
24.82
MDL number:
UNSPSC Code:
12352200

form

powder

density

2.29 g/mL at 25 °C (lit.)

SMILES string

B#N

InChI

1S/BN/c1-2

InChI key

PZNSFCLAULLKQX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Extremely thin layers of boron nitrides (BN) have emerged as an important class of non-graphene based 2D-materials which find applications in flexible & printed electronics, LEDs, and sensors.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flexible and Transparent MoS2 Field-Effect Transistors on Hexagonal Boron Nitride-Graphene Heterostructures.
Leet GH, et al.
ACS Nano, 7(9), 7931-7936 (2013)
F Withers et al.
Nature materials, 14(3), 301-306 (2015-02-03)
The advent of graphene and related 2D materials has recently led to a new technology: heterostructures based on these atomically thin crystals. The paradigm proved itself extremely versatile and led to rapid demonstration of tunnelling diodes with negative differential resistance
Per Haberkant et al.
Biochimica et biophysica acta, 1841(8), 1022-1030 (2014-01-21)
Understanding biological processes at the mechanistic level requires a systematic charting of the physical and functional links between all cellular components. While protein-protein and protein-nucleic acid networks have been subject to many global surveys, other critical cellular components such as
In vivo profiling and visualization of cellular protein-lipid interactions using bifunctional fatty acids.
Per Haberkant et al.
Angewandte Chemie (International ed. in English), 52(14), 4033-4038 (2013-03-02)

Articles

Advances in scalable synthesis and processing of two-dimensional materials

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service