Skip to Content
Merck
All Photos(1)

Key Documents

900388

Sigma-Aldrich

Inulin-13C

(from chicory), ≥97 atom % 13C, ≥97% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
R$21,791.00

R$21,791.00


Check Cart for Availability


Select a Size

Change View
100 MG
R$21,791.00

About This Item

Linear Formula:
13C6nH10n+2O5n+1
UNSPSC Code:
12352005
NACRES:
NA.13

R$21,791.00


Check Cart for Availability

Quality Level

biological source

(from chicory)

isotopic purity

≥97 atom % 13C

Assay

≥97% (CP)

form

powder

mass shift

M+6 (per repeating unit)

Related Categories

Packaging

This product is available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service