Skip to Content
Merck
All Photos(1)

Key Documents

900375

Sigma-Aldrich

α-Tocopherol-(trimethylphenyl-13C9)

≥99 atom % 13C, ≥96% (CP)

Synonym(s):

Vitamin E-(trimethylphenyl-13C9)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
R$8,947.00

R$8,947.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 MG
R$8,947.00

About This Item

Empirical Formula (Hill Notation):
13C9C20H50O2
Molecular Weight:
439.64
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

R$8,947.00


Check Cart for Availability

Request more information

isotopic purity

≥99 atom % 13C

Assay

≥96% (CP)

form

liquid

technique(s)

mass spectrometry (MS): suitable

color

yellow to brown

mass shift

M+9

storage temp.

−20°C

SMILES string

CC(C)CCC[C@@H](C)CCC[C@@H](C)CCCC1(C)CC[13C]2=[13C]([13CH3])[13C](O)=[13C]([13CH3])[13C]([13CH3])=[13C]2O1

InChI

1S/C29H50O2/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(30)23(5)24(6)28(26)31-29/h20-22,30H,9-19H2,1-8H3/t21-,22-,29?/m1/s1/i5+1,6+1,7+1,23+1,24+1,25+1,26+1,27+1,28+1

InChI key

GVJHHUAWPYXKBD-OPIPRMLOSA-N

Related Categories

General description

α-Tocopherol (vitamin E) is a lipid-soluble antioxidant found in whole grains, nuts, and oils. It is known to scavenge reactive oxygen species by interacting with lipids during oxidative stress.[1][2]
α-Tocopherol-(trimethylphenyl-13C9) is an isotope of vitamin E wherein all the six phenyl carbons and three methyl carbons attached to the phenyl ring are replaced by 13C6 carbon.

Application

α-Tocopherol-(trimethylphenyl-13C9) can be used as a stable isotope internal standard for data interpretation in specific biological samples.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

ANTIOXIDANTS-Role of Antioxidant Nutrients in Defense Systems
Encyclopedia of food sciences and nutrition, 282-289 (2003)
The role of alpha-tocopherol in plant stress tolerance
Munne-Bosch S
Journal of Plant Physiology, 162(7), 743-748 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service