Skip to Content
Merck
All Photos(1)

Key Documents

900190

Sigma-Aldrich

Silver nanoparticle inkjet ink

Synonym(s):

Ag ink, Conductive ink, Silver ink, Xerox® silver nanoparticle inkjet ink, xcm-nsIJ-RS1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

description

Conductivity: ≥1×105 Scm-1
surface tension 28 - 30 mN/m at 25°C

form

dispersion

viscosity

4.0-6.0 cP (shear viscosity at 25°C, 1-400 s-1)

cation traces

Ag: 43-45 wt. % (by ash)

storage temp.

2-8°C

Other Notes

Caution, do not freeze.

Legal Information

Xerox is a registered trademark of Xerox Corporation

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Asp. Tox. 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

205.9 °F

Flash Point(C)

96.6 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service